Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929352 | LOC101929352 | Anti-sense | Chr. 10 - 31187883 - 31261552 on NCBI Build GRCh38 | ||
| Downstream | ZEB1 antisense RNA 1 | ZEB1-AS1 | 127.2kb | Sense | Chr. 10 - 31316528 - 31319095 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map