Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105376480 | LOC105376480 | -202.8kb | Anti-sense | Chr. 10 - 30723219 - 30723740 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101929352 | LOC101929352 | 260.7kb | Anti-sense | Chr. 10 - 31187883 - 31261552 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map