Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927066 | LOC101927066 | Sense | Chr. 8 - 96951864 - 97446497 on NCBI Build GRCh38 | ||
| Downstream | TSPY like 5 | TSPYL5 | 31.3kb | Sense | Chr. 8 - 97273486 - 97277948 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map