Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | monoamine oxidase B | MAOB | Sense | Chr. X - 43766610 - 43882475 on NCBI Build GRCh38 | ||
| Downstream |
NDP, norrin cystine knot growth factor |
NDP |
EVR2, FEVR, ND |
110.5kb | Sense | Chr. X - 43948776 - 43973675 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map