Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gap junction protein beta 2 | GJB2 |
CX26, DFNA3, DFNA3A, DFNB1, DFNB1A, HID, KID, NSRD1, PPK |
-1.4kb | Anti-sense | Chr. 13 - 20187463 - 20192975 on NCBI Build GRCh38 |
| Downstream | gap junction protein beta 6 | GJB6 |
CX30, DFNA3, DFNA3B, DFNB1B, ECTD2, ED2, EDH, HED, HED2 |
26.0kb | Anti-sense | Chr. 13 - 20221962 - 20232395 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map