Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929371 | LOC101929371 | -14.9kb | Sense | Chr. 20 - 9986333 - 10007116 on NCBI Build GRCh38 | |
| Downstream | SNAP25 antisense RNA 1 | SNAP25-AS1 | 441.0 | Anti-sense | Chr. 20 - 10023812 - 10219506 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map