Assay Details
| Gene Symbol: GABPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323906.1 | NP_001310835.1 | 10 | 2558 | 105 | |
| NM_001323911.1 | NP_001310840.1 | 9 | 2444 | 105 | ||
| GenBank mRNA | AF143872.1 | - | 1 | 546 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11 |
MLLT11 | AF1Q | -56.1kb | Sense | Chr. 1 - 151059675 - 151068497 on NCBI Build GRCh38 |
| Downstream | semaphorin 6C | SEMA6C |
SEMAY, m-SemaY, m-SemaY2 |
6.1kb | Anti-sense | Chr. 1 - 151131685 - 151148408 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GABPB2
Genomic Map