Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dynein axonemal heavy chain 10 | DNAH10 | -32.4kb | Anti-sense | Chr. 12 - 123761995 - 123935720 on NCBI Build GRCh38 | |
| Downstream | zinc finger protein 664 | ZNF664 |
ZFOC1, ZNF176 |
4.7kb | Anti-sense | Chr. 12 - 123973215 - 124015439 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map