Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 34 | CCDC34 |
L15, NY-REN-41, RAMA3 |
-105.1kb | Anti-sense | Chr. 11 - 27338514 - 27363248 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105376671 | LOC105376671 | lnc-BBOX1-1 | 3.5kb | Sense | Chr. 11 - 27471731 - 27482433 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map