Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1 |
HECW1 | NEDL1 | -10.5kb | Sense | Chr. 7 - 43112580 - 43566001 on NCBI Build GRCh38 |
| Downstream | serine/threonine kinase 17a | STK17A | DRAK1 | 7.6kb | Sense | Chr. 7 - 43583093 - 43627379 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map