Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TMC3 antisense RNA 1 | TMC3-AS1 | -406.2kb | Anti-sense | Chr. 15 - 81324333 - 81443076 on NCBI Build GRCh38 | |
| Downstream | mex-3 RNA binding family member B | MEX3B |
MEX-3B, RKHD3, RNF195 |
191.0kb | Sense | Chr. 15 - 82041778 - 82046143 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map