Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928778 | LOC101928778 | -213.4kb | Anti-sense | Chr. 1 - 177700524 - 177710330 on NCBI Build GRCh38 | |
| Downstream |
SEC16 homolog B, endoplasmic reticulum export factor |
SEC16B |
LZTR2, PGPR-p117, RGPR, SEC16S |
4.8kb | Sense | Chr. 1 - 177929107 - 177969915 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map