Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UHRF1 binding protein 1 | UHRF1BP1 |
C6orf107, ICBP90, dJ349A12.1 |
Sense | Chr. 6 - 34792017 - 34877514 on NCBI Build GRCh38 | |
| Downstream | TATA-box binding protein associated factor 11 | TAF11 |
MGC:15243, PRO2134, TAF2I, TAFII28 |
83.8kb | Anti-sense | Chr. 6 - 34877778 - 34888071 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map