Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DAN domain BMP antagonist family member 5 | DAND5 |
CER2, CERL2, CKTSF1B3, COCO, CRL2, DANTE, GREM3, SP1 |
-107.2kb | Sense | Chr. 19 - 12969618 - 12974753 on NCBI Build GRCh38 |
| Downstream |
LYL1, basic helix-loop-helix family member |
LYL1 | bHLHa18 | 14.6kb | Anti-sense | Chr. 19 - 13099028 - 13103160 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map