Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HIG1 hypoxia inducible domain family member 1A | HIGD1A |
HIG1, RCF1a |
-39.1kb | Sense | Chr. 3 - 42782908 - 42804535 on NCBI Build GRCh38 |
| Downstream | cytochrome P450 family 8 subfamily B member 1 | CYP8B1 |
CP8B, CYP12 |
27.9kb | Sense | Chr. 3 - 42872192 - 42876141 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map