Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger SWIM-type containing 4 | ZSWIM4 | Anti-sense | Chr. 19 - 13795424 - 13832254 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC284454 | LOC284454 | 37.6kb | Sense | Chr. 19 - 13834516 - 13836289 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map