Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serine/arginine-rich splicing factor 3 pseudogene | LOC100500773 | -111.9kb | Sense | Chr. 8 - 95946984 - 95948348 on NCBI Build GRCh38 | |
| Downstream | growth differentiation factor 6 | GDF6 |
BMP-13, BMP13, CDMP2, KFM, KFS, KFS1, KFSL, LCA17, MCOP4, MCOPCB6, SCDO4, SGM1 |
81.9kb | Sense | Chr. 8 - 96142330 - 96160792 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map