Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 88C | CCDC88C |
DAPLE, HKRP2, KIAA1509, SCA40 |
Sense | Chr. 14 - 91271323 - 91418498 on NCBI Build GRCh38 | |
| Downstream | protein phosphatase 4 regulatory subunit 3A | PPP4R3A |
FLFL1, KIAA2010, MSTP033, PP4R3, PP4R3A, SMEK1, smk-1, smk1 |
72.9kb | Sense | Chr. 14 - 91457481 - 91510554 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map