Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928583 | LOC101928583 | -62.5kb | Sense | Chr. 3 - 170467285 - 170755205 on NCBI Build GRCh38 | |
| Downstream | ribosomal protein L22 like 1 | RPL22L1 | 139.9kb | Anti-sense | Chr. 3 - 170864875 - 170870256 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map