Assay Details
| Gene Symbol: FUBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011519172.2 | XP_011517474.1 | 17 | 2130 | 95 | |
| GenBank mRNA | BC128537.1 | - | 1 | 755 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6856 | MIR6856 | hsa-mir-6856 | -7.0kb | Sense | Chr. 9 - 130626297 - 130626363 on NCBI Build GRCh38 |
| Downstream | PR domain 12 | PRDM12 |
HSAN8, PFM9 |
30.0kb | Sense | Chr. 9 - 130664594 - 130682997 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

FUBP3
Genomic Map