Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WD repeat domain 83 | WDR83 | MORG1 | Anti-sense | Chr. 19 - 12666804 - 12675832 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105372280 | LOC105372280 | 11.9kb | Anti-sense | Chr. 19 - 12687998 - 12688420 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map