Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Yip1 domain family member 5 | YIPF5 |
FinGER5, SB140, SMAP-5, SMAP5, YIP1A |
-183.8kb | Sense | Chr. 5 - 144158159 - 144170714 on NCBI Build GRCh38 |
| Downstream | PRELI domain containing 2 | PRELID2 | 981.3kb | Sense | Chr. 5 - 145587325 - 145835369 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map