Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 8061 | MIR8061 | hsa-mir-8061 | Anti-sense | Chr. 19 - 54645309 - 54645383 on NCBI Build GRCh38 | |
| Downstream | leukocyte immunoglobulin-like receptor pseudogene 2 | LILRP2 |
CD85m, ILT10, LILRA5 |
44.8kb | Anti-sense | Chr. 19 - 54707916 - 54713452 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map