Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tweety family member 1 | TTYH1 | Anti-sense | Chr. 19 - 54415431 - 54436719 on NCBI Build GRCh38 | ||
| Downstream | LENG8 antisense RNA 1 | LENG8-AS1 | 25.5kb | Sense | Chr. 19 - 54444816 - 54449038 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map