Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serpin family B member 7 | SERPINB7 |
MEGSIN, PPKN, TP55 |
-66.3kb | Anti-sense | Chr. 18 - 63753047 - 63805376 on NCBI Build GRCh38 |
| Downstream | serpin family B member 10 | SERPINB10 |
PI-10, PI10 |
17.9kb | Anti-sense | Chr. 18 - 63907969 - 63936111 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map