Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serpin family B member 7 | SERPINB7 |
MEGSIN, PPKN, TP55 |
-97.5kb | Anti-sense | Chr. 18 - 63753047 - 63805376 on NCBI Build GRCh38 |
| Downstream | histocompatibility minor serpin domain containing | HMSD |
ACC-6, ACC6, C18orf53, HSMD-v |
41.2kb | Anti-sense | Chr. 18 - 63949259 - 63969329 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map