Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MIER family member 3 | MIER3 | Anti-sense | Chr. 5 - 56919602 - 56952130 on NCBI Build GRCh38 | ||
| Downstream | GC-rich promoter binding protein 1 | GPBP1 |
GPBP, SSH6, VASCULIN |
214.3kb | Sense | Chr. 5 - 57173826 - 57264679 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map