Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serpin family B member 11 (gene/pseudogene) | SERPINB11 |
EPIPIN, SERPIN11 |
Sense | Chr. 18 - 63702304 - 63723893 on NCBI Build GRCh38 | |
| Downstream | serpin family B member 7 | SERPINB7 |
MEGSIN, PPKN, TP55 |
37.7kb | Sense | Chr. 18 - 63753047 - 63805376 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map