Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | F-box protein 7 | FBXO7 |
FBX, FBX07, FBX7, PARK15, PKPS |
-307.4kb | Anti-sense | Chr. 22 - 32474682 - 32498831 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105373006 | LOC105373006 | 302.2kb | Anti-sense | Chr. 22 - 33108529 - 33116294 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map