Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | growth/differentiation factor 3 | LOC105378979 | -72.5kb | Anti-sense | Chr. 5 - 56766395 - 56772157 on NCBI Build GRCh38 | |
| Downstream | SET domain containing 9 | SETD9 | C5orf35 | 63.5kb | Sense | Chr. 5 - 56909260 - 56928900 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map