Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FOXP4 antisense RNA 1 | FOXP4-AS1 | -632.0 | Anti-sense | Chr. 6 - 41523895 - 41548621 on NCBI Build GRCh38 | |
| Downstream | microRNA 4641 | MIR4641 | 49.3kb | Sense | Chr. 6 - 41598723 - 41598788 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map