Assay Details
| Gene Symbol: LOC107986762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012908.1 | XP_016868397.1 | 1 | 67 | ||
| XM_017012909.1 | XP_016868398.1 | 2 | 67 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fascin actin-bundling protein 1 | FSCN1 |
FAN1, HSN, SNL, p55 |
-9.3kb | Sense | Chr. 7 - 5592805 - 5606656 on NCBI Build GRCh38 |
| Downstream | ring finger protein 216 | RNF216 |
CAHH, TRIAD3, U7I1, UBCE7IP1, ZIN |
3.9kb | Anti-sense | Chr. 7 - 5620041 - 5781730 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017012908.1, XM_017012909.1 transcripts.
Set Membership: |
|

LOC107986762
Genomic Map