Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 29b-1 | MIR29B1 |
MIRN29B1, miR-29b, miRNA29B1, mir-29b-1 |
-9.5kb | Anti-sense | Chr. 7 - 130877459 - 130877539 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100506860 | LOC100506860 | 26.2kb | Sense | Chr. 7 - 130913464 - 130921946 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map