Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ST7 overlapping transcript 3 | ST7-OT3 |
NCRNA00026, ST7, ST7OT3 |
Anti-sense | Chr. 7 - 117182681 - 117209937 on NCBI Build GRCh38 | |
| Downstream | Wnt family member 2 | WNT2 |
INT1L1, IRP |
76.2kb | Sense | Chr. 7 - 117276631 - 117323289 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map