Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ALOX12 antisense RNA 1 | ALOX12-AS1 | Anti-sense | Chr. 17 - 6985123 - 7012334 on NCBI Build GRCh38 | ||
| Downstream | ribonuclease K | RNASEK | 13.5kb | Sense | Chr. 17 - 7012417 - 7014533 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map