Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mediator complex subunit 12 like | MED12L |
NOPAR, TNRC11L, TRALP, TRALPUSH |
Sense | Chr. 3 - 151085665 - 151436677 on NCBI Build GRCh38 | |
| Downstream | G protein-coupled receptor 87 | GPR87 |
FKSG78, GPR95, KPG_002 |
12.8kb | Anti-sense | Chr. 3 - 151294086 - 151316848 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map