Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | colon adenocarcinoma hypermethylated (non-protein coding) | CAHM | LINC00468 | -50.3kb | Anti-sense | Chr. 6 - 163413065 - 163413950 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC102724152 | LOC102724152 | 198.2kb | Anti-sense | Chr. 6 - 163664366 - 163671484 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map