Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 2 member 10 | SLC2A10 |
ATS, GLUT10 |
-398.9kb | Sense | Chr. 20 - 46708358 - 46736347 on NCBI Build GRCh38 |
| Downstream | microRNA 3616 | MIR3616 | mir-3616 | 30.2kb | Sense | Chr. 20 - 47166967 - 47167058 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map