Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4744 | MIR4744 | -142.4kb | Sense | Chr. 18 - 49049687 - 49049768 on NCBI Build GRCh38 | |
| Downstream | chromosome 18 open reading frame 32 | C18orf32 | 287.0kb | Sense | Chr. 18 - 49481178 - 49487274 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map