Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | muscle associated receptor tyrosine kinase | MUSK |
CMS9, FADS |
-222.1kb | Anti-sense | Chr. 9 - 110668189 - 110806633 on NCBI Build GRCh38 |
| Downstream | microRNA 7702 | MIR7702 | hsa-mir-7702 | 240.2kb | Sense | Chr. 9 - 111271156 - 111271214 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map