Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927943 | LOC101927943 | -211.7kb | Anti-sense | Chr. 18 - 44676927 - 44679717 on NCBI Build GRCh38 | |
| Downstream | microRNA 4319 | MIR4319 | 75.1kb | Anti-sense | Chr. 18 - 44970082 - 44970166 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map