Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WAC antisense RNA 1 (head to head) | WAC-AS1 | -69.4kb | Anti-sense | Chr. 10 - 28519917 - 28532354 on NCBI Build GRCh38 | |
| Downstream | BMP and activin membrane bound inhibitor | BAMBI | NMA | 73.6kb | Sense | Chr. 10 - 28677495 - 28682939 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map