Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100505795 | LOC100505795 | -53.8kb | Anti-sense | Chr. 1 - 164769116 - 164774641 on NCBI Build GRCh38 | |
| Downstream | LIM homeobox transcription factor 1 alpha | LMX1A |
LMX1, LMX1.1 |
371.9kb | Anti-sense | Chr. 1 - 165201867 - 165356715 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map