Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small lysine rich protein 1 | SMKR1 | -238.7kb | Sense | Chr. 7 - 129502479 - 129512932 on NCBI Build GRCh38 | |
| Downstream | microRNA 182 | MIR182 |
MIRN182, miRNA182, mir-182 |
15.9kb | Anti-sense | Chr. 7 - 129770383 - 129770492 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map