Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G protein-coupled receptor 15 | GPR15 | BOB | -49.4kb | Anti-sense | Chr. 3 - 98531899 - 98534679 on NCBI Build GRCh38 |
| Downstream | ST3GAL6 antisense RNA 1 | ST3GAL6-AS1 | 130.0kb | Sense | Chr. 3 - 98714333 - 98732651 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map