Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UBE2E2 antisense RNA 1 (head to head) | UBE2E2-AS1 | -235.6kb | Anti-sense | Chr. 3 - 23195070 - 23202993 on NCBI Build GRCh38 | |
| Downstream | UBE2E1 antisense RNA 1 | UBE2E1-AS1 | 363.2kb | Anti-sense | Chr. 3 - 23804024 - 23806905 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map