Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DLGAP1 antisense RNA 4 | DLGAP1-AS4 | -147.1kb | Anti-sense | Chr. 18 - 3962353 - 4013943 on NCBI Build GRCh38 | |
| Downstream | DLGAP1 antisense RNA 5 | DLGAP1-AS5 | 102.0kb | Anti-sense | Chr. 18 - 4264602 - 4296000 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map