Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tubulin beta 6 class V | TUBB6 |
HsT1601, TUBB-5 |
-11.1kb | Anti-sense | Chr. 18 - 12307669 - 12329826 on NCBI Build GRCh38 |
| Downstream | PRELI domain containing 3A | PRELID3A |
C18orf43, HFL-EDDG1, SLMO1 |
64.8kb | Anti-sense | Chr. 18 - 12407896 - 12432238 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map