Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DDB1 and CUL4 associated factor 17 | DCAF17 | C2orf37 | -54.8kb | Sense | Chr. 2 - 171434166 - 171491029 on NCBI Build GRCh38 |
| Downstream | dynein cytoplasmic 1 intermediate chain 2 | DYNC1I2 |
DIC74, DNCI2, IC2 |
138.8kb | Sense | Chr. 2 - 171687409 - 171750158 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map