Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 25 member 29 | SLC25A29 |
C14orf69, CACL, ORNT3 |
Sense | Chr. 14 - 100277986 - 100306547 on NCBI Build GRCh38 | |
| Downstream | microRNA 345 | MIR345 |
MIRN345, hsa-mir-345, mir-345 |
2.4kb | Anti-sense | Chr. 14 - 100307859 - 100307956 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map